N-(2-Chloroethyl)-N'-(4-fluorophenyl)urea

Supplier: Apollo Scientific
Warning

PC8779-1G PC8779-5G
APOSPC8779-1GEA 94.5 EUR
APOSPC8779-1G APOSPC8779-5G
N-(2-Chloroethyl)-N'-(4-fluorophenyl)urea
N-(2-Chloroethyl)-N'-(4-fluorophenyl)urea
Formula: C₉H₁₀ClFN₂O
Melting Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD03787786
CAS Number: 13908-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR