3,4,5-Trifluorobenzylamine 98%

Supplier: Apollo Scientific
Danger

Synonyms: (3,4,5-Trifluorophenyl)methanamine

PC8792-1G PC8792-25G PC8792-5G PC8792-100G
APOSPC8792-1GEA 0 EUR
APOSPC8792-1G APOSPC8792-25G APOSPC8792-5G APOSPC8792-100G
3,4,5-Trifluorobenzylamine 98%
3,4,5-Trifluorobenzylamine
Formula: C₇H₆F₃N
MW: 161,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236318
CAS Number: 235088-69-4
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR