3-Chloro-5-(trifluoromethyl)phenylboronic acid

Supplier: Apollo Scientific
Warning

Synonyms: (3-Chloro-5-(trifluoromethyl)phenyl)boronic acid

PC8834-1G PC8834-25G PC8834-5G
APOSPC8834-1GEA 21.9 EUR
APOSPC8834-1G APOSPC8834-25G APOSPC8834-5G
3-Chloro-5-(trifluoromethyl)phenylboronic acid
3-Chloro-5-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BClF₃O₂
MW: 224,3700 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07778020
CAS Number: 1160561-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR