4-Bromo-3,5-difluorophenol
Supplier: Apollo Scientific
|
Warning
|
PC8836-1G
PC8836-5G
PC8836-25G
APOSPC8836-1GEA
43.7
EUR
APOSPC8836-1G
APOSPC8836-5G
APOSPC8836-25G
4-Bromo-3,5-difluorophenol
4-Bromo-3,5-difluorophenol
Formula:
C₆H₃BrF₂O Melting Pt: 73…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD08062385 CAS Number: 130191-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...