4-Bromo-2-nitro-1-(trifluoromethyl)benzene

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-2-nitrobenzotrifluoride

PC8938-10G PC8938-1G PC8938-5G
APOSPC8938-10GEA 96 EUR
APOSPC8938-10G APOSPC8938-1G APOSPC8938-5G
4-Bromo-2-nitro-1-(trifluoromethyl)benzene
4-Bromo-2-nitro-1-(trifluoromethyl)benzene
Formula: C₇H₃BrF₃NO₂
MW: 270,0000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226284
CAS Number: 251115-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR