2,3,5,6-Tetrafluorobenzoic acid

Supplier: Apollo Scientific
Danger

PC8975-25G PC8975-5G
APOSPC8975-25GEA 36.4 EUR
APOSPC8975-25G APOSPC8975-5G
2,3,5,6-Tetrafluorobenzoic acid
2,3,5,6-Tetrafluorobenzoic acid
Formula: C₇H₂F₄O₂
MW: 194,09 g/mol
Melting Pt: 148…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002408
CAS Number: 652-18-6
EINECS: 435-890-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR