4-Iodo-2-(trifluoromethyl)pyridine 95%

Supplier: Apollo Scientific
Warning

PC8981-1G PC8981-250MG PC8981-5G
APOSPC8981-1GEA 101 EUR
APOSPC8981-1G APOSPC8981-250MG APOSPC8981-5G
4-Iodo-2-(trifluoromethyl)pyridine 95%
4-Iodo-2-(trifluoromethyl)pyridine
Formula: C₆H₃F₃IN
Melting Pt: 24…26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08234939
CAS Number: 590371-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR