2-Fluoro-4-methylanisole 95%

Supplier: Apollo Scientific

PC900018-1G PC900018-5G
APOSPC900018-1GEA 139 EUR
APOSPC900018-1G APOSPC900018-5G
2-Fluoro-4-methylanisole 95%
2-Fluoro-4-methylanisole
Formula: C₈H₉FO
MW: 140,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060270
CAS Number: 399-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR