8-Fluoro-2-quinolinol 98%

Supplier: Apollo Scientific

Synonyms: 8-Fluoroquinolin-2(1H)-one

PC900635-1G PC900635-5G
APOSPC900635-1GEA 59.6 EUR
APOSPC900635-1G APOSPC900635-5G
8-Fluoro-2-quinolinol 98%
8-Fluoro-2-quinolinol
Formula: C₉H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD12026113
CAS Number: 71738-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR