3-Fluoro-4-(1-pyrrolidinyl)aniline 98%

Supplier: Apollo Scientific

PC901082-1G PC901082-5G PC901082-10G
APOSPC901082-1GEA 168 EUR
APOSPC901082-1G APOSPC901082-5G APOSPC901082-10G
3-Fluoro-4-(1-pyrrolidinyl)aniline 98%
3-Fluoro-4-(1-pyrrolidinyl)aniline
Formula: C₁₀H₁₃FN₂
MW: 180,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07403468
CAS Number: 93246-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR