4-Fluoro-N,N-dimethylbenzamide 97%
Supplier: Apollo Scientific
Synonyms:
N,N-Dimethyl 4-fluorobenzamide
PC901456-25G
PC901456-5G
APOSPC901456-25GEA
426
EUR
APOSPC901456-25G
APOSPC901456-5G
4-Fluoro-N,N-dimethylbenzamide 97%
4-Fluoro-N,N-dimethylbenzamide
Formula:
C₉H₁₀FNO MW: 167,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01214152 CAS Number: 24167-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...