Methyl-4-fluoro-2-methylbenzoate 98%

Supplier: Apollo Scientific

Synonyms: Methyl 4-fluoro-2-methylbenzoate

PC901494-100G PC901494-25G
APOSPC901494-100GEA 245 EUR
APOSPC901494-100G APOSPC901494-25G
Methyl-4-fluoro-2-methylbenzoate 98%
Methyl 4-fluoro-2-methylbenzoate
Formula: C₉H₉FO₂
MW: 168,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203790
CAS Number: 174403-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR