4-Fluoro-N-tert-butylbenzamide 98%

Supplier: Apollo Scientific

Synonyms: N-t-Butyl-4-fluorobenzamide

PC901495-25G PC901495-5G
APOSPC901495-25GEA 435 EUR
APOSPC901495-25G APOSPC901495-5G
4-Fluoro-N-tert-butylbenzamide 98%
4-Fluoro-N-tert-butylbenzamide
Formula: C₁₁H₁₄FNO
MW: 195,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463095
CAS Number: 49834-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR