5-Bromo-8-fluoroquinoline 98%

Supplier: Apollo Scientific
Warning

PC901861-1G PC901861-250MG PC901861-5G
APOSPC901861-1GEA 72.5 EUR
APOSPC901861-1G APOSPC901861-250MG APOSPC901861-5G
5-Bromo-8-fluoroquinoline 98%
5-Bromo-8-fluoroquinoline
Formula: C₉H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD11855895
CAS Number: 1133115-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR