3-Fluoro-4-thiocyanatoaniline 98%

Supplier: Apollo Scientific

PC901928-1G PC901928-5G
APOSPC901928-1GEA 147 EUR
APOSPC901928-1G APOSPC901928-5G
3-Fluoro-4-thiocyanatoaniline 98%
3-Fluoro-4-thiocyanatoaniline
Formula: C₇H₅FN₂S
Storage Temperature: Ambient
MDL Number: MFCD11855824
CAS Number: 14512-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR