6-Fluoro-8-methoxyquinoline 97%

Supplier: Apollo Scientific

PC901942-1G PC901942-5G
APOSPC901942-1GEA 219 EUR
APOSPC901942-1G APOSPC901942-5G
6-Fluoro-8-methoxyquinoline 97%
6-Fluoro-8-methoxyquinoline
Formula: C₁₀H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD15143547
CAS Number: 887769-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR