3-Chloro-2,6-difluoropyridine 95%
Supplier: Apollo Scientific
PC902115-1G
PC902115-5G
APOSPC902115-1GEA
132
EUR
APOSPC902115-1G
APOSPC902115-5G
3-Chloro-2,6-difluoropyridine 95%
3-Chloro-2,6-difluoropyridine
Formula:
C₅H₂ClF₂N Storage Temperature: Ambient |
MDL Number:
MFCD11044285 CAS Number: 52208-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...