6,8-Difluoroquinoline 98%
Supplier: Apollo Scientific
PC902177-10G
PC902177-1G
PC902177-5G
APOSPC902177-10GEA
502
EUR
APOSPC902177-10G
APOSPC902177-1G
APOSPC902177-5G
6,8-Difluoroquinoline 98%
6,8-Difluoroquinoline
Formula:
C₉H₅F₂N MW: 165,1400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03662296 CAS Number: 145241-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...