4-Fluoro-2-mercaptobenzothiazole 98%
Supplier: Apollo Scientific
Synonyms:
4-Fluorobenzo[d]thiazole-2(3H)-thione
PC902198-1G
PC902198-5G
APOSPC902198-1GEA
85.8
EUR
APOSPC902198-1G
APOSPC902198-5G
4-Fluoro-2-mercaptobenzothiazole 98%
4-Fluoro-2-mercaptobenzothiazole
Formula:
C₇H₄FNS₂ MW: 185,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06408877 CAS Number: 154327-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...