6-(Trifluoromethyl)isatin 98%

Supplier: Apollo Scientific

PC902245-1G PC902245-5G PC902245-10G
APOSPC902245-1GEA 114 EUR
APOSPC902245-1G APOSPC902245-5G APOSPC902245-10G
6-(Trifluoromethyl)isatin 98%
6-(Trifluoromethyl)isatin
Formula: C₉H₄F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD08457887
CAS Number: 343-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR