3-Fluorophthalonitrile 95%

Supplier: Apollo Scientific

PC902673-25G PC902673-5G
APOSPC902673-25GEA 1250 EUR
APOSPC902673-25G APOSPC902673-5G
3-Fluorophthalonitrile 95%
3-Fluorophthalonitrile
Formula: C8H3FN2
MW: 146,1212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052458
CAS Number: 65610-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR