2,2,3,3-Tetrafluoropropyl-p-toluenesulphonate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,2,3,3-Tetrafluoropropyl 4-methylbenzenesulphonate
PC9037-100G
PC9037-25G
PC9037-5G
APOSPC9037-100GEA
364
EUR
APOSPC9037-100G
APOSPC9037-25G
APOSPC9037-5G
2,2,3,3-Tetrafluoropropyl-p-toluenesulphonate
2,2,3,3-Tetrafluoropropyl-p-toluenesulphonate
Formula:
C₁₀H₁₀F₄O₃S MW: 286,25 g/mol Boiling Pt: 134…136 °C/0,5mmHg Melting Pt: 13…16 °C Density: 1,397 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00155960 CAS Number: 786-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...