2,3,4,6-Tetrafluorobenzyl alcohol 95%

Supplier: Apollo Scientific

PC904651-1G PC904651-250MG
APOSPC904651-1GEA 342 EUR
APOSPC904651-1G APOSPC904651-250MG
2,3,4,6-Tetrafluorobenzyl alcohol 95%
2,3,4,6-Tetrafluorobenzyl alcohol
Formula: C₇H₄F₄O
Storage Temperature: Ambient
MDL Number: MFCD03701105
CAS Number: 53001-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR