3-(2-Fluorophenyl)-3-oxopropanenitrile 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Fluorobenzoylacetonitrile

PC9054-1G PC9054-5G
APOSPC9054-1GEA 62.5 EUR
APOSPC9054-1G APOSPC9054-5G
3-(2-Fluorophenyl)-3-oxopropanenitrile 97%
3-(2-Fluorophenyl)-3-oxopropanenitrile
Formula: C₉H₆FNO
MW: 163,15 g/mol
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD02260800
CAS Number: 31915-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR