3-Fluorobenzoylacetonitrile 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-(3-Fluorophenyl)-3-oxopropanenitrile
PC9056-1G
PC9056-25G
PC9056-5G
APOSPC9056-1GEA
130
EUR
APOSPC9056-1G
APOSPC9056-25G
APOSPC9056-5G
3-Fluorobenzoylacetonitrile 98%
3-Fluorobenzoylacetonitrile
Formula:
C₉H₆FNO MW: 163,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02260777 CAS Number: 21667-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...