Methyl-3-fluoro-2-thiophenecarboxylate 95%

Supplier: Apollo Scientific

Synonyms: Methyl 3-fluorothiophene-2-carboxylate

PC906382-5G
APOSPC906382-5GEA 127 EUR
APOSPC906382-5G
Methyl-3-fluoro-2-thiophenecarboxylate 95%
Methyl-3-fluoro-2-thiophenecarboxylate
Formula: C₆H₅FO₂S
MW: 160,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16606491
CAS Number: 100421-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR