2,4-Difluoro-3-methylbenzylamine 95%
Supplier: Apollo Scientific
PC906439-1G
PC906439-5G
APOSPC906439-1GEA
248
EUR
APOSPC906439-1G
APOSPC906439-5G
2,4-Difluoro-3-methylbenzylamine 95%
2,4-Difluoro-3-methylbenzylamine
Formula:
C₈H₉F₂N MW: 157,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08275293 CAS Number: 847502-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...