6-Fluoro-2-methyl-2,3-dihydro-1H-inden-1-one 95%

Supplier: Apollo Scientific

Synonyms: 6-Fluoro-2-methylindanone

PC906461-100MG PC906461-250MG PC906461-500MG
APOSPC906461-100MGEA 205 EUR
APOSPC906461-100MG APOSPC906461-250MG APOSPC906461-500MG
6-Fluoro-2-methyl-2,3-dihydro-1H-inden-1-one 95%
6-Fluoro-2-methyl-2,3-dihydro-1H-inden-1-one
Formula: C₁₀H₉FO
MW: 164,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00029129
CAS Number: 37794-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR