4-Amino-5-fluoro-2-methoxypyrimidine 95%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Fluoro-2-methoxypyrimidin-4-amine

PC906580-1G PC906580-5G
APOSPC906580-1GEA 154 EUR
APOSPC906580-1G APOSPC906580-5G
4-Amino-5-fluoro-2-methoxypyrimidine 95%
4-Amino-5-fluoro-2-methoxypyrimidine
Formula: C₅H₆FN₃O
MW: 143,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264289
CAS Number: 1993-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR