4'-Methoxy-2,2,2-trifluoroacetophenone 95%

Supplier: Apollo Scientific
Warning

PC907828-10G PC907828-25G PC907828-5G PC907828-1G
APOSPC907828-10GEA 0 EUR
APOSPC907828-10G APOSPC907828-25G APOSPC907828-5G APOSPC907828-1G
4'-Methoxy-2,2,2-trifluoroacetophenone 95%
4'-Methoxy-2,2,2-trifluoroacetophenone
Formula: C₉H₇F₃O₂
Density: 1,32 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00018061
CAS Number: 711-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR