6-Fluoro-3,4-dihydroquinolin-2(1H)-one 95%

Supplier: Apollo Scientific

PC907895-1G PC907895-250MG
APOSPC907895-1GEA 597 EUR
APOSPC907895-1G APOSPC907895-250MG
6-Fluoro-3,4-dihydroquinolin-2(1H)-one 95%
6-Fluoro-3,4-dihydroquinolin-2(1H)-one
Formula: C₉H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD09971532
CAS Number: 75893-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR