5-(3-Fluorophenyl)-1,3,4-oxadiazol-2-amine 95%
Supplier: Apollo Scientific
PC907909-1G
PC907909-250MG
APOSPC907909-1GEA
85.8
EUR
APOSPC907909-1G
APOSPC907909-250MG
5-(3-Fluorophenyl)-1,3,4-oxadiazol-2-amine 95%
5-(3-Fluorophenyl)-1,3,4-oxadiazol-2-amine
Formula:
C₈H₆FN₃O Storage Temperature: Ambient |
MDL Number:
MFCD09028454 CAS Number: 341978-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...