3-(Trifluoroacetyl)pyridine 95%

Supplier: Apollo Scientific

Synonyms: 2,2,2-Trifluoro-1-(pyridin-3-yl)ethanol

PC908420-1G PC908420-250MG
APOSPC908420-1GEA 806 EUR
APOSPC908420-1G APOSPC908420-250MG
3-(Trifluoroacetyl)pyridine 95%
3-(Trifluoroacetyl)pyridine
Formula: C₇H₆F₃NO
Storage Temperature: Ambient
MDL Number: MFCD16067922
CAS Number: 138624-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR