N-Methyl-3-(trifluoromethyl)benzylamine 95%
Supplier: Apollo Scientific
PC908684-1G
PC908684-5G
APOSPC908684-1GEA
71.3
EUR
APOSPC908684-1G
APOSPC908684-5G
N-Methyl-3-(trifluoromethyl)benzylamine 95%
N-Methyl-3-(trifluoromethyl)benzylamine
Formula:
C₉H₁₀F₃N MW: 189,181 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115410 CAS Number: 90390-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...