5-Fluoroisoindoline hydrochloride 97%
Supplier: Apollo Scientific
PC908748-1G
PC908748-5G
APOSPC908748-1GEA
159
EUR
APOSPC908748-1G
APOSPC908748-5G
5-Fluoroisoindoline hydrochloride 97%
5-Fluoroisoindoline hydrochloride
Formula:
C8H9ClFN Storage Temperature: Ambient |
MDL Number:
MFCD09832541 CAS Number: 685565-15-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...