1-(4-Fluorophenyl)-2-methylpropylamine 96%
Supplier: Apollo Scientific
PC908877-100MG
PC908877-1G
PC908877-250MG
APOSPC908877-100MGEA
168
EUR
APOSPC908877-100MG
APOSPC908877-1G
APOSPC908877-250MG
1-(4-Fluorophenyl)-2-methylpropylamine 96%
1-(4-Fluorophenyl)-2-methylpropylamine
Formula:
C₁₀H₁₄FN MW: 167,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07339047 CAS Number: 863668-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...