4-(2-Fluorophenyl)but-3-en-2-one 97%
Supplier: Apollo Scientific
PC910901-1G
PC910901-5G
APOSPC910901-1GEA
209
EUR
APOSPC910901-1G
APOSPC910901-5G
4-(2-Fluorophenyl)but-3-en-2-one 97%
4-(2-Fluorophenyl)but-3-en-2-one
Formula:
C₁₀H₉FO MW: 164,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01221416 CAS Number: 2143-80-8 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...