5-(Trifluoromethyl)picolinic acid 97%
Supplier: Apollo Scientific
|
Warning
|
PC9132-1G
PC9132-5G
APOSPC9132-1GEA
21.9
EUR
APOSPC9132-1G
APOSPC9132-5G
5-(Trifluoromethyl)picolinic acid 97%
5-(Trifluoromethyl)picolinic acid
Formula:
C₇H₄F₃NO₂ Melting Pt: 133…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD04113632 CAS Number: 80194-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...