2-Chloro-1,1,1,3,4,4,4-heptafluorobutane

Supplier: Apollo Scientific

PC9137-25G PC9137-5G
APOSPC9137-25GEA 379 EUR
APOSPC9137-25G APOSPC9137-5G
2-Chloro-1,1,1,3,4,4,4-heptafluorobutane
2-Chloro-1,1,1,3,4,4,4-heptafluorobutane
Formula: C₄H₂ClF₇
Storage Temperature: Ambient
MDL Number: MFCD03094541
CAS Number: 507453-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR