2-Chloro-5-fluoronicotinic acid 96%

Supplier: Apollo Scientific
Warning

PC9179-5G PC9179-25G PC9179-100G
APOSPC9179-5GEA 14.7 EUR
APOSPC9179-5G APOSPC9179-25G APOSPC9179-100G
2-Chloro-5-fluoronicotinic acid 96%
2-Chloro-5-fluoronicotinic acid
Formula: C₆H₃ClFNO₂
MW: 175,5449 g/mol
Melting Pt: 141…144 °C
Storage Temperature: Ambient
MDL Number: MFCD03092932
CAS Number: 38186-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR