2-Fluoro-3-nitropyridine
Supplier: Apollo Scientific
|
Warning
|
PC9190-100G
PC9190-25G
APOSPC9190-100GEA
358
EUR
APOSPC9190-100G
APOSPC9190-25G
2-Fluoro-3-nitropyridine
2-Fluoro-3-nitropyridine
Formula:
C₅H₃FN₂O₂ Boiling Pt: 109…110 °C/10mmHg Melting Pt: 17,8 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095068 CAS Number: 1480-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...