4-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde 97%

Supplier: Apollo Scientific
Warning

PC9200-1G PC9200-5G
APOSPC9200-1GEA 14.7 EUR
APOSPC9200-1G APOSPC9200-5G
4-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde 97%
4-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde
Formula: C₁₄H₁₀ClFO₂
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00052227
CAS Number: 172932-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR