2,3,5-Trifluorobenzyl alcohol 97%

Supplier: Apollo Scientific
Warning

PC9241-25G
APOSPC9241-25GEA 0 EUR
APOSPC9241-25G
2,3,5-Trifluorobenzyl alcohol 97%
2,3,5-Trifluorobenzyl alcohol
Formula: C₇H₅F₃O
MW: 162,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083542
CAS Number: 67640-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR