Pentafluorophenyl sulphide 95%
Supplier: Apollo Scientific
|
Warning
|
PC9256-1G
PC9256-5G
PC9256-25G
APOSPC9256-1GEA
29
EUR
APOSPC9256-1G
APOSPC9256-5G
APOSPC9256-25G
Pentafluorophenyl sulphide 95%
Pentafluorophenyl sulphide
Formula:
C₁₂F₁₀S Melting Pt: 86…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000293 CAS Number: 1043-50-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...