Triethyloxonium hexafluorophosphate

Supplier: Apollo Scientific
Danger

PC9262-25G PC9262-5G
APOSPC9262-25GEA 799 EUR
APOSPC9262-25G APOSPC9262-5G
Triethyloxonium hexafluorophosphate
Triethyloxonium hexafluorophosphate
Formula: C₆H₁₅F₆OP
MW: 189,99 g/mol
Melting Pt: 145 °C
Flash Pt: 57 °C
Storage Temperature: Refrigerator
CAS Number: 17950-40-2
EINECS: 241-876-6
UN: 2925
ADR: 4.1+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR