4-Iodo-3-(trifluoromethyl)benzonitrile 95%

Supplier: Apollo Scientific
Warning

PC9307-25G PC9307-5G
APOSPC9307-25GEA 509 EUR
APOSPC9307-25G APOSPC9307-5G
4-Iodo-3-(trifluoromethyl)benzonitrile 95%
4-Iodo-3-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₃IN
MW: 297,0158 g/mol
Melting Pt: 92…99 °C
Storage Temperature: Ambient
MDL Number: MFCD08458134
CAS Number: 161320-00-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR