2,3,5-Trifluorobenzaldehyde 98%

Supplier: Apollo Scientific
Warning

PC9324-25G PC9324-5G PC9324-10G
APOSPC9324-25GEA 201 EUR
APOSPC9324-25G APOSPC9324-5G APOSPC9324-10G
2,3,5-Trifluorobenzaldehyde 98%
2,3,5-Trifluorobenzaldehyde
Formula: C₇H₃F₃O
MW: 160,1 g/mol
Boiling Pt: 165 °C
Density: 1,407 g/cm³ (25 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00083538
CAS Number: 126202-23-1
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR