3-(4-Fluoro-2-nitrophenoxy)-2-thiophenecarboxylic acid 97%

Supplier: Apollo Scientific

PC9346-10G PC9346-1G PC9346-5G
APOSPC9346-10GEA 800 EUR
APOSPC9346-10G APOSPC9346-1G APOSPC9346-5G
3-(4-Fluoro-2-nitrophenoxy)-2-thiophenecarboxylic acid 97%
3-(4-Fluoro-2-nitrophenoxy)-2-thiophenecarboxylic acid
Formula: C₁₁H₆FNO₅S
MW: 283,24 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00214992
CAS Number: 303152-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR