2-(4-Fluorophenoxy)ethanimidamide hydrochloride
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(4-Fluorophenoxy)acetamidine hydrochloride
PC9348-1G
PC9348-250MG
APOSPC9348-1GEA
321
EUR
APOSPC9348-1G
APOSPC9348-250MG
2-(4-Fluorophenoxy)ethanimidamide hydrochloride
2-(4-Fluorophenoxy)ethanimidamide hydrochloride
Formula:
C₈H₁₀ClFN₂O MW: 204,63 g/mol Melting Pt: 156…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD01114620 CAS Number: 318951-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...