5-Fluoro-1H-benzimidazole 98%
Supplier: Apollo Scientific
Synonyms:
5-Fluoro-1H-benzo[d]imidazole
PC9393-1G
PC9393-5G
APOSPC9393-1GEA
96
EUR
APOSPC9393-1G
APOSPC9393-5G
5-Fluoro-1H-benzimidazole 98%
5-Fluoro-1H-benzimidazole
Formula:
C₇H₅FN₂ MW: 136,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00792438 CAS Number: 1977-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...